Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002181.1 | NP_001002181.1 | 7 - 8 | 1030 | 101 | |
GenBank mRNA | BC074043.1 | - | 7 - 8 | 1030 | 101 | |
BC155262.1 | - | 7 - 8 | 969 | 101 | ||
DQ100356.1 | - | 7 - 8 | 965 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |