Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002198.1 | NP_001002198.1 | 4 - 5 | 992 | 75 | |
XM_021480593.1 | XP_021336268.1 | 4 - 5 | 1059 | 75 | ||
GenBank mRNA | BC074031.1 | - | 4 - 5 | 992 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |