Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gadd45ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002216.1 | NP_001002216.1 | 1 - 2 | 158 | 68 | |
GenBank mRNA | BC074023.1 | - | 1 - 2 | 130 | 68 | |
BC075932.1 | - | 1 - 2 | 158 | 68 | ||
BC152637.1 | - | 1 - 2 | 150 | 68 | ||
BC164910.1 | - | 1 - 2 | 77 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |