Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aqp10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002349.1 | NP_001002349.1 | 1 - 2 | 160 | 83 | |
GenBank mRNA | BC075911.1 | - | 1 - 2 | 160 | 83 | |
FJ655388.1 | - | 1 - 2 | 128 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Inventoried Probe spans exons |