Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bpnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002354.1 | NP_001002354.1 | 6 - 7 | 540 | 66 | |
XM_021468396.1 | XP_021324071.1 | 7 - 8 | 652 | 66 | ||
GenBank mRNA | BC075903.1 | - | 6 - 7 | 540 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |