Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: galm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002373.1 | NP_001002373.1 | 3 - 4 | 613 | 91 | |
XM_021480558.1 | XP_021336233.1 | 3 - 4 | 91 | |||
GenBank mRNA | BC076039.1 | - | 3 - 4 | 613 | 91 | |
BC153585.1 | - | 3 - 4 | 606 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |