Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: entpd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002379.2 | NP_001002379.2 | 4 - 5 | 496 | 86 | |
GenBank mRNA | BC075878.1 | - | 4 - 5 | 481 | 86 | |
DT068903.1 | - | 4 - 5 | 481 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |