Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002436.2 | NP_001002436.2 | 1 - 2 | 319 | 74 | |
GenBank mRNA | AI415997.1 | - | 1 - 2 | 301 | 74 | |
BC076129.1 | - | 1 - 2 | 285 | 74 | ||
BC096994.1 | - | 1 - 2 | 311 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |