Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpy30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_130111.1 | 5 - 6 | 425 | 117 | ||
GenBank mRNA | BC076125.1 | - | 5 - 6 | 379 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |