Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crygn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007784.1 | NP_001007785.1 | 2 - 3 | 307 | 67 | |
GenBank mRNA | AY738747.1 | - | 2 - 3 | 307 | 67 | |
BC097242.1 | - | 2 - 3 | 321 | 67 | ||
BC121745.1 | - | 2 - 3 | 321 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |