Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drgx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037105.1 | NP_001032182.1 | 4 - 5 | 429 | 62 | |
XM_005156850.4 | XP_005156907.1 | 5 - 6 | 1027 | 62 | ||
GenBank mRNA | AY753211.1 | - | 4 - 5 | 429 | 62 | |
BC162787.1 | - | 4 - 5 | 459 | 62 | ||
BC162791.1 | - | 4 - 5 | 459 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |