Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc8a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039144.1 | NP_001034233.1 | 4 - 5 | 2185 | 84 | |
GenBank mRNA | AY934776.1 | - | 4 - 5 | 2185 | 84 | |
BC163419.1 | - | 4 - 5 | 2212 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |