Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casp6b.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039980.1 | NP_001035069.1 | 2 - 3 | 224 | 118 | |
XM_017354587.2 | XP_017210076.1 | 1 - 2 | 205 | 118 | ||
GenBank mRNA | BC114318.1 | - | 2 - 3 | 224 | 118 | |
MG957998.1 | - | 2 - 3 | 201 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |