Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003200513.4 | XP_003200561.1 | 3 - 4 | 753 | 118 | |
GenBank mRNA | BC065891.1 | - | 3 - 4 | 671 | 118 | |
BC115235.1 | - | 3 - 4 | 714 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |