Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: top1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044324.2 | NP_001037789.1 | 16 - 17 | 1803 | 82 | |
GenBank mRNA | BC117588.1 | - | 16 - 17 | 1803 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |