Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rad51ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044338.2 | NP_001037803.2 | 3 - 4 | 427 | 116 | |
XM_009299961.3 | XP_009298236.1 | 3 - 4 | 441 | 116 | ||
GenBank mRNA | BC133972.1 | - | 3 - 4 | 427 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |