Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044816.1 | NP_001038281.1 | 3 - 4 | 378 | 84 | |
XM_009306499.2 | XP_009304774.1 | 5 - 6 | 1183 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |