Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cort | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045431.2 | NP_001038896.1 | 1 - 2 | 208 | 118 | |
GenBank mRNA | BC122388.1 | - | 1 - 2 | 209 | 118 | |
BI472739.1 | - | 1 - 2 | 212 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |