Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppm1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320183.1 | NP_001307112.1 | 2 - 3 | 795 | 58 | |
GenBank mRNA | BC124282.1 | - | 2 - 3 | 609 | 58 | |
BC164393.1 | - | 2 - 3 | 573 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |