Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076619.1 | NP_001070087.1 | 5 - 6 | 1314 | 139 | |
XM_017356912.2 | XP_017212401.1 | 5 - 6 | 1430 | 139 | ||
GenBank mRNA | BC124441.1 | - | 5 - 6 | 1314 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |