Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uhmk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076659.1 | NP_001070127.1 | 2 - 3 | 907 | 94 | |
GenBank mRNA | BC124286.1 | - | 2 - 3 | 907 | 94 | |
BC164787.1 | - | 2 - 3 | 564 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |