Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uhmk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076659.1 | NP_001070127.1 | 4 - 5 | 1201 | 99 | |
GenBank mRNA | BC124286.1 | - | 4 - 5 | 1201 | 99 | |
BC164787.1 | - | 4 - 5 | 858 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |