Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: esm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076741.1 | NP_001070209.1 | 2 - 3 | 579 | 72 | |
GenBank mRNA | BC124217.1 | - | 2 - 3 | 579 | 72 | |
BC164957.1 | - | 2 - 3 | 487 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |