Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr5a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077272.2 | NP_001070740.2 | 6 - 7 | 1491 | 84 | |
NM_214779.2 | NP_999944.2 | 6 - 7 | 1496 | 84 | ||
GenBank mRNA | BC124756.1 | - | 6 - 7 | 1462 | 84 | |
BC163909.1 | - | 6 - 7 | 1459 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |