Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2t | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077295.2 | NP_001070763.1 | 6 - 7 | 544 | 99 | |
GenBank mRNA | AW420785.1 | - | 1 - 2 | 243 | 99 | |
BC124718.1 | - | 6 - 7 | 545 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |