Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agmat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077551.1 | NP_001071019.1 | 3 - 4 | 699 | 69 | |
GenBank mRNA | BC124326.1 | - | 3 - 4 | 699 | 69 | |
BC164767.1 | - | 3 - 4 | 602 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |