Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phkg1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077612.1 | NP_001071080.1 | 2 - 3 | 243 | 77 | |
XM_021481442.1 | XP_021337117.1 | 2 - 3 | 77 | |||
GenBank mRNA | BC125847.1 | - | 2 - 3 | 243 | 77 | |
BC164489.1 | - | 1 - 2 | 111 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |