Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmprss4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077738.1 | NP_001071206.1 | 3 - 4 | 223 | 76 | |
XM_005157490.4 | XP_005157547.1 | 3 - 4 | 506 | 76 | ||
GenBank mRNA | BC125834.1 | - | 3 - 4 | 223 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |