Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079944.1 | NP_001073413.1 | 4 - 5 | 771 | 152 | |
GenBank mRNA | BC142737.1 | - | 4 - 5 | 769 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |