Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081692.2 | NP_001075161.2 | 15 - 16 | 2052 | 78 | |
XM_005160248.4 | XP_005160305.1 | 15 - 16 | 2010 | 78 | ||
XM_009293357.3 | XP_009291632.1 | 15 - 16 | 2638 | 78 | ||
XM_009293365.3 | XP_009291640.1 | 15 - 16 | 1939 | 78 | ||
GenBank mRNA | BC128874.1 | - | 6 - 7 | 848 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |