Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ldlrap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080635.1 | NP_001074104.1 | 7 - 8 | 922 | 63 | |
XM_021480671.1 | XP_021336346.1 | 7 - 8 | 946 | 63 | ||
GenBank mRNA | BC129425.1 | - | 7 - 8 | 922 | 63 | |
BC164254.1 | - | 7 - 8 | 770 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |