Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slco1f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080666.2 | NP_001074135.2 | 12 - 13 | 1753 | 94 | |
GenBank mRNA | BC129232.1 | - | 12 - 13 | 1723 | 94 | |
KP294319.1 | - | 11 - 12 | 1626 | 94 | ||
KP294320.1 | - | 11 - 12 | 1626 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |