Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dachd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080673.1 | NP_001074142.1 | 8 - 9 | 1626 | 72 | |
XM_005167749.4 | XP_005167806.1 | 7 - 8 | 1618 | 72 | ||
XM_005167750.4 | XP_005167807.1 | 7 - 8 | 1621 | 72 | ||
GenBank mRNA | BC129504.1 | - | 8 - 9 | 1626 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |