Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080687.1 | NP_001074156.1 | 6 - 7 | 849 | 85 | |
GenBank mRNA | BC129485.1 | - | 6 - 7 | 849 | 85 | |
BC154814.1 | - | 6 - 7 | 797 | 85 | ||
BC164099.1 | - | 6 - 7 | 826 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |