Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx3xa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126423.1 | NP_001119895.1 | 10 - 11 | 1296 | 87 | |
XM_005167718.4 | XP_005167775.1 | 11 - 12 | 1400 | 87 | ||
XM_005167720.4 | XP_005167777.1 | 9 - 10 | 1292 | 87 | ||
XM_009304751.3 | XP_009303026.1 | 10 - 11 | 1337 | 87 | ||
XM_017357705.2 | XP_017213194.1 | 11 - 12 | 1397 | 87 | ||
XM_017357706.2 | XP_017213195.1 | 11 - 12 | 1397 | 87 | ||
XM_021478726.1 | XP_021334401.1 | 13 - 14 | 1737 | 87 | ||
XM_021478727.1 | XP_021334402.1 | 13 - 14 | 1734 | 87 | ||
XM_021478728.1 | XP_021334403.1 | 13 - 14 | 1734 | 87 | ||
XM_021478729.1 | XP_021334404.1 | 12 - 13 | 1689 | 87 | ||
XM_021478730.1 | XP_021334405.1 | 12 - 13 | 1677 | 87 | ||
XM_021478731.1 | XP_021334406.1 | 12 - 13 | 1653 | 87 | ||
XM_021478733.1 | XP_021334408.1 | 14 - 15 | 1650 | 87 | ||
XM_021478734.1 | XP_021334409.1 | 13 - 14 | 1590 | 87 | ||
XM_021478735.1 | XP_021334410.1 | 13 - 14 | 1566 | 87 | ||
XM_021478736.1 | XP_021334411.1 | 10 - 11 | 1352 | 87 | ||
GenBank mRNA | BC133162.1 | - | 10 - 11 | 1296 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |