Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grhl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083072.1 | NP_001076541.1 | 9 - 10 | 1255 | 91 | |
XM_005159580.4 | XP_005159637.1 | 10 - 11 | 1552 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |