Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taar11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083077.1 | NP_001076546.1 | 1 | 852 | 70 | |
XM_021468526.1 | XP_021324201.1 | 2 | 894 | 70 | ||
GenBank mRNA | BC163651.1 | - | 1 | 938 | 70 | |
BC163664.1 | - | 1 | 938 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |