Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfcq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083099.1 | NP_001076568.1 | 1 - 2 | 109 | 118 | |
XM_009293696.2 | XP_009291971.1 | 1 - 2 | 246 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Danio rerio Probe spans exons |