Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfcq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083099.1 | NP_001076568.1 | 3 - 4 | 1208 | 95 | |
XM_009293696.2 | XP_009291971.1 | 3 - 4 | 1345 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Danio rerio Probe spans exons |