Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083552.1 | NP_001077021.1 | 3 - 4 | 289 | 85 | |
GenBank mRNA | BC133920.1 | - | 3 - 4 | 289 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |