Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sfrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083571.1 | NP_001077040.1 | 1 - 2 | 599 | 92 | |
GenBank mRNA | BC133945.1 | - | 1 - 2 | 599 | 92 | |
BC153379.1 | - | 1 - 2 | 507 | 92 | ||
BC154248.1 | - | 1 - 2 | 601 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |