Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slco1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348086.1 | NP_001335015.1 | 5 - 6 | 609 | 137 | |
XM_021474933.1 | XP_021330608.1 | 6 - 7 | 627 | 137 | ||
GenBank mRNA | BC139695.1 | - | 5 - 6 | 636 | 137 | |
KP294314.1 | - | 5 - 6 | 600 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |