Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089485.1 | NP_001082954.1 | 4 - 5 | 578 | 77 | |
XM_021466574.1 | XP_021322249.1 | 4 - 5 | 565 | 77 | ||
GenBank mRNA | BC135028.1 | - | 4 - 5 | 578 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |