Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tprg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089528.2 | NP_001082997.1 | 3 - 4 | 430 | 65 | |
GenBank mRNA | BC139672.1 | - | 3 - 4 | 431 | 65 | |
BC158178.1 | - | 3 - 4 | 431 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |