Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtf3ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089544.2 | NP_001083013.2 | 2 - 3 | 342 | 71 | |
GenBank mRNA | BC133956.1 | - | 2 - 3 | 270 | 71 | |
EH546778.1 | - | 2 - 3 | 311 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |