Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhcga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089577.1 | NP_001083046.1 | 9 - 10 | 1371 | 81 | |
GenBank mRNA | AB286867.1 | - | 9 - 10 | 1396 | 81 | |
BC139524.1 | - | 9 - 10 | 1371 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |