Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp4r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110371.1 | NP_001103841.1 | 4 - 5 | 472 | 107 | |
GenBank mRNA | BC129134.1 | - | 4 - 5 | 456 | 107 | |
BC154497.1 | - | 4 - 5 | 472 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |