Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: igsf21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110473.1 | NP_001103943.1 | 1 - 2 | 243 | 125 | |
GenBank mRNA | BC134916.1 | - | 1 - 2 | 128 | 125 | |
CU458945.1 | - | 1 - 2 | 273 | 125 | ||
KM655674.1 | - | 1 - 2 | 211 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |