Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110837.1 | NP_001104307.1 | 1 - 2 | 249 | 93 | |
GenBank mRNA | BC162934.1 | - | 1 - 2 | 252 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |