Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifnphi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111083.1 | NP_001104553.1 | 2 - 3 | 212 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |